Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Sepabeads™ SP850

Catalog No. 111003855 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3855 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3855 Supplier MilliporeSigma™ Supelco™ Supplier No. 13599
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Swelling in toluene is 19%

Sepabeads™ SP850 is a macroporous polymeric hydrophobic resin of styrene-divinylbenzene. This resin can be used as receiving phase in passive samplers. This resin rapidly adsorbs okadaic acid and dinophysistoxins.
TRUSTED_SUSTAINABILITY

Specifications

Pore Size 38 Å
Packaging Bucket
Quantity 1000 g
Surface Area (Metric) ˜1000 m2/g
Product Line Sepabeads
Density 1.01 g/mL at 25°C (true wet)(lit.)

Sepabeads is a registered trademark of Mitsubishi Chemical Corp.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.