Learn More
Medchemexpress LLC HY-10194 10mg Medchemexpress, Sepantronium (bromide) CAS:781661-94-7 Purity:>98%
SDP
Catalog No. 502028690
Encompass_Preferred
Medchemexpress, HY-10194 10mg Sepantronium (bromide) CAS:781661-94-7 Sepantronium bromide (YM-155) is a survivin inhibitor with an IC50 of 0.54 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8690 Supplier Medchemexpress LLC Supplier No. HY1019410MG
Medchemexpress, HY-10194 10mg Sepantronium (bromide) CAS:781661-94-7 Sepantronium bromide (YM-155) is a survivin inhibitor with an IC50 of 0.54 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.