Learn More
Medchemexpress LLC Sepantronium (bromide) | 781600-92-9 | >98% | 750.56 | 25MG
SDP
Catalog No. 5000347685
Encompass_Preferred
Sepantronium bromide (YM155) is a small molecule inhibitor of survivin, a member of the inhibitor of apoptosis protein (IAP) family. It exhibits potent anticancer activity by suppressing survivin expression, leading to apoptosis in various cancer cell lines.
  • Small molecule inhibitor of survivin
  • IC50 of 0.54 nM in a cell-free assay
  • Potent anticancer activity
  • Suppresses survivin expression
  • Leads to apoptosis in various cancer cell lines
  • Under clinical investigation for the treatment of various cancers
Catalog No. 50-003-47685 Supplier Medchemexpress LLC Supplier No. 25MGHY10194
Add to Cart
Add to Cart
Sepantronium bromide (YM155) is a small molecule inhibitor of survivin, a member of the inhibitor of apoptosis protein (IAP) family. It exhibits potent anticancer activity by suppressing survivin expression, leading to apoptosis in various cancer cell lines.
  • Small molecule inhibitor of survivin
  • IC50 of 0.54 nM in a cell-free assay
  • Potent anticancer activity
  • Suppresses survivin expression
  • Leads to apoptosis in various cancer cell lines
  • Under clinical investigation for the treatment of various cancers