Learn More
Medchemexpress LLC Sepantronium bromide | 781661-94-7 | MFCD11983133 | 98.9% | 443.3 g/mol | C20H19BrN4O3 | 100 MG
SDP
Catalog No. 5000248753
Encompass_Preferred
Sepantronium bromide (YM-155) is a small-molecule survivin inhibitor used in research to suppress survivin expression and study apoptosis in cancer models. It shows potent activity in cellular assays and is supplied at high purity for biochemical and cell-based experiments.
  • Potent survivin inhibition (IC50 ~0.54 nM).
  • High purity: 98.9%.
  • Available as a solid or as a 10 mM solution for flexible dosing.
  • Chemical formula C20H19BrN4O3; mol wt 443.3 g/mol.
  • Intended for research use only.
Catalog No. 50-002-48753 Supplier Medchemexpress LLC Supplier No. HY10194100MG
Sepantronium bromide (YM-155) is a small-molecule survivin inhibitor used in research to suppress survivin expression and study apoptosis in cancer models. It shows potent activity in cellular assays and is supplied at high purity for biochemical and cell-based experiments.
  • Potent survivin inhibition (IC50 ~0.54 nM).
  • High purity: 98.9%.
  • Available as a solid or as a 10 mM solution for flexible dosing.
  • Chemical formula C20H19BrN4O3; mol wt 443.3 g/mol.
  • Intended for research use only.