Learn More
Bioworld Separating Gel Buffer (4X), 500 mL
Catalog No. 50196665
Encompass_Preferred
Product DescriptionSeparating gel buffer has a concentration of 4X and contains 1.5M Tris at a pH value of 8.8 and 0.4% SDS. Final concentrations of Tris and SDS after dilution is 0.375M and 0.1% respectively. Product is stored at room temperature.Proteomics grade.
Catalog No. 50-196-665 Supplier Bioworld Supplier No. 105300083
Product DescriptionSeparating gel buffer has a concentration of 4X and contains 1.5M Tris at a pH value of 8.8 and 0.4% SDS. Final concentrations of Tris and SDS after dilution is 0.375M and 0.1% respectively. Product is stored at room temperature.Proteomics grade.