Learn More
Chemglass Life Sciences 20L Globe Shaped Separatory Funnel, Large Capacity, #45/50 Outer Stopper Neck, 10mm PTFE Stpk
SDP
Catalog No. NC1270311
Encompass_Preferred
Funnel, Separatory, 22000mL, Globe Shaped, 10mmPTFE Stopcock, 45/50 Stopper
Catalog No. NC1270311 Supplier Chemglass Life Sciences Supplier No. CG173908
Funnel, Separatory, 22000mL, Globe Shaped, 10mmPTFE Stopcock, 45/50 Stopper