Learn More
Ace Glass, Inc. Funnel, separatory, PTFE stopcock, PTFE stopper, 2L
SDP
Catalog No. NC0649863
Encompass_Preferred
2L Squibb separatory funnel, featuring a 1:5 PTFE stopcock plug with a 6mm plug bore, and a standard taper PTFE stopper.
Catalog No. NC0649863 Supplier Ace Glass, Inc. Supplier No. 722872
2L Squibb separatory funnel, featuring a 1:5 PTFE stopcock plug with a 6mm plug bore, and a standard taper PTFE stopper.