Learn More
Chemglass Life Sciences Chemglass PYREX Squibb separatory funnel, 125 mL, PTFE stopcock, stopper #22
SDP
Catalog No. NC1907475
Encompass_Preferred
This separatory funnel is a pear-shaped PYREX glass funnel designed for routine liquid-liquid separations in the laboratory. It is fitted with a PTFE stopcock and a stopper to provide secure phase transfer and controlled dispensing.
  • Capacity: 125 mL
  • Material: PYREX glass
  • Stopcock type: PTFE stopcock
  • Stopper size: #22
  • Style: Squibb (pear-shaped) separatory funnel
  • Typical use: liquid-liquid extraction and phase separation
Catalog No. NC1907475 Supplier Chemglass Life Sciences Supplier No. CG8626125
This separatory funnel is a pear-shaped PYREX glass funnel designed for routine liquid-liquid separations in the laboratory. It is fitted with a PTFE stopcock and a stopper to provide secure phase transfer and controlled dispensing.
  • Capacity: 125 mL
  • Material: PYREX glass
  • Stopcock type: PTFE stopcock
  • Stopper size: #22
  • Style: Squibb (pear-shaped) separatory funnel
  • Typical use: liquid-liquid extraction and phase separation