Learn More
Chemglass Life Sciences Chemglass PYREX Squibb Separatory Funnel, 250 mL, PTFE stopcock, #22 HDPE stopper
SDP
Catalog No. NC1907477
Encompass_Preferred
PYREX Squibb separatory funnel designed for laboratory liquid-liquid extractions and phase separations. The funnel combines a corrosion-resistant PTFE stopcock with a closed-bottom HDPE stopper and a pear-shaped body to promote efficient layer separation and reusable performance.
  • 250 mL capacity.
  • Body material: glass.
  • PTFE stopcock for chemically resistant flow control.
  • Closed-bottom high-density polyethylene stopper, size 22.
  • Pear-shaped (Squibb) body for efficient phase separation.
  • Reusable glass construction with plastic components.
Catalog No. NC1907477 Supplier Chemglass Life Sciences Supplier No. CG8626250
Add to Cart
Add to Cart
PYREX Squibb separatory funnel designed for laboratory liquid-liquid extractions and phase separations. The funnel combines a corrosion-resistant PTFE stopcock with a closed-bottom HDPE stopper and a pear-shaped body to promote efficient layer separation and reusable performance.
  • 250 mL capacity.
  • Body material: glass.
  • PTFE stopcock for chemically resistant flow control.
  • Closed-bottom high-density polyethylene stopper, size 22.
  • Pear-shaped (Squibb) body for efficient phase separation.
  • Reusable glass construction with plastic components.