Learn More
Bioworld SEPAROPOREAGAROSE) 4B
Catalog No. 50197740
Encompass
A high gel strength bioagarose matrix for gel filtration and coupling of affinity ligands suitable for analytical and preparative protein purifications. Larger volumes available at more competitive prices(enquire). Specifications: Parti...
Catalog No. 50-197-740 Supplier Bioworld Supplier No. 201810311
A high gel strength bioagarose matrix for gel filtration and coupling of affinity ligands suitable for analytical and preparative protein purifications. Larger volumes available at more competitive prices(enquire). Specifications: Parti...