Learn More
Bioworld SEPAROPOREAGAROSE) 4B-CL
Supplier:  Bioworld 201810323
Encompass
A cross-linked bioagarose medium for gel filtration and coupling of affinity ligands. Hydrophilic backbone results in low non-specific interactions and high protein recoveries . Provides excellent separation of proteins over a wide molec...
Catalog No. 50-197-745