Learn More
Bioworld SEPAROPOREAGAROSE) 4B-MB
Catalog No. 50197747
Encompass
SEPAROPORE (AGAROSE) 4B-MB (MACROBEADS): An agarose bead matrix for gel filtration and coupling of affinity ligands suitable for analytical and preparative protein purifications. Specifications Particle size (range): 150-265m; Matrix: Ag...
Catalog No. 50-197-747 Supplier Bioworld Supplier No. 201810331
SEPAROPORE (AGAROSE) 4B-MB (MACROBEADS): An agarose bead matrix for gel filtration and coupling of affinity ligands suitable for analytical and preparative protein purifications. Specifications Particle size (range): 150-265m; Matrix: Ag...