Learn More
Bioworld SEPAROPOREAGAROSE) 6B
Catalog No. 50197773
Encompass
A high gel strength bioagarose matrix for gel filtration and coupling of affinity ligands suitable for analytical and preparative protein purifications. Larger volumes available at more competitive prices(enquie). Specifications: Parti...
Catalog No. 50-197-773 Supplier Bioworld Supplier No. 201810431
A high gel strength bioagarose matrix for gel filtration and coupling of affinity ligands suitable for analytical and preparative protein purifications. Larger volumes available at more competitive prices(enquie). Specifications: Parti...