Learn More
Bioworld SEPAROPOREAGAROSE) 6B-CL
Supplier:  Bioworld 201810363
Encompass
A cross-linked high strength agarose medium for gel filtration and coupling of affinity ligands. A hydrophilic backbone results in low non specific interactions and high yields. Excellent separation over a wide molecular weight range. S...
Catalog No. 50-197-755