Learn More
Bioworld SEPAROPOREAGAROSE) 6B-MB
Supplier:  Bioworld 201810352
Encompass
An agarose bead matrix for gel filtration and coupling of affinity ligands suitable for analytical and preparative protein purifications. Specifications Particle size (range): 150-265m; Matrix: Agarose 6% beads; pH stability: 4-9. Usage...
Catalog No. 50-197-751