Learn More
Sepax Technologies Inc Sepax Generik FPLC Empty Column AF, 15 x 250 mm, Glass
SDP
Catalog No. 502337060
Encompass
FPLC empty column has 3 configurations to choose from: AF(one fixed endpiece and one adjustable endpiece), AA(two adjustable endpieces), and FF(non-adjustable with two fixed endpieces). The versatile glass empty columns are ideal for FPLC purification based on following characters: high pressure tolerance, up to 900 psi (or 60 bar); easy use two adjustable endpieces configuration; wide range of inner diameter, from 6.6 mm to 50 mm with various lengths; and it is compatible with most solvents used in FPLC purification
Catalog No. 50-233-7060 Supplier Sepax Technologies Inc Supplier No. 2020001525AF
FPLC empty column has 3 configurations to choose from: AF(one fixed endpiece and one adjustable endpiece), AA(two adjustable endpieces), and FF(non-adjustable with two fixed endpieces). The versatile glass empty columns are ideal for FPLC purification based on following characters: high pressure tolerance, up to 900 psi (or 60 bar); easy use two adjustable endpieces configuration; wide range of inner diameter, from 6.6 mm to 50 mm with various lengths; and it is compatible with most solvents used in FPLC purification