Learn More
Sepax Technologies Inc Sepax HPLC Accessories Column coupler with 10cm length flexible tubing, high pressure 10-32 fitting with 1/16 ODPEEK tubing. Recommended to connect analytical columns in series
SDP
Catalog No. 502337441
Encompass
Sepax HPLC Accessories Column coupler with 10cm length flexible tubing, high pressure 10-32 fitting with 1/16 ODPEEK tubing. Recommended to connect analytical columns in series
Catalog No. 50-233-7441 Supplier Sepax Technologies Inc Supplier No. 102006COUPLER
Sepax HPLC Accessories Column coupler with 10cm length flexible tubing, high pressure 10-32 fitting with 1/16 ODPEEK tubing. Recommended to connect analytical columns in series