Learn More
Sepax Technologies Inc Sepax HPLC Accessories One-piece PEEK coupler, recommended to connect columns in series.
SDP
Catalog No. 502336714
Encompass
Sepax HPLC Accessories One-piece PEEK coupler, recommended to connect columns in series.
Catalog No. 50-233-6714 Supplier Sepax Technologies Inc Supplier No. 102002COUPLER
Sepax HPLC Accessories One-piece PEEK coupler, recommended to connect columns in series.