Learn More
Sepax Technologies Inc Sepax HPLC Column Bio-C4, 5 µm, 300 Å, 30 x 100 mm
SDP
Catalog No. 502338318
Encompass
Monomeric and fully endcapped Bio-C4 packing is bonded with butyl groups that lead to moderate hydrophobicity. Bio-C4 packings of 300 Å pore size selection are ideal for high resolution mapping of peptides and separation of intact proteins, and natural and synthetic peptides.
Catalog No. 50-233-8318 Supplier Sepax Technologies Inc Supplier No. 11004530010
Monomeric and fully endcapped Bio-C4 packing is bonded with butyl groups that lead to moderate hydrophobicity. Bio-C4 packings of 300 Å pore size selection are ideal for high resolution mapping of peptides and separation of intact proteins, and natural and synthetic peptides.