Learn More
Sepax Technologies Inc Sepax HPLC Column Bio-C8, Guard cartridge with holder, 5 µm, 300 Å, 2 x 10 mm
SDP
Catalog No. 502337812
Encompass
Bio-C8 phase is made of monomeric and fully endcapped chemistry. The uniform stationary phase allows the separation to achieve high selectivity and high efficiency. Bio-C8 packings of 300 Å pore size selection are ideal for high resolution mapping of peptides and separation of natural and synthetic peptides and small proteins.
Catalog No. 50-233-7812 Supplier Sepax Technologies Inc Supplier No. 1080852001C
Bio-C8 phase is made of monomeric and fully endcapped chemistry. The uniform stationary phase allows the separation to achieve high selectivity and high efficiency. Bio-C8 packings of 300 Å pore size selection are ideal for high resolution mapping of peptides and separation of natural and synthetic peptides and small proteins.