Learn More
Sepax Technologies Inc Sepax HPLC Column BioService Affinity HSA, PEEK, 2.1 x 50 mm
SDP
Catalog No. 502336788
Encompass
Sepax offers an off-the-shelf product range of affinity columns conjugated with Human Serum Albumin (HSA) for our customers who are looking to asset mAb Immunogen Cross-Reactivity. The columns available can be easily attached to standard HPLC/UPLC instruments and provide an unique solution for process monitoring. For customers interested in custom immobilization contact us for more information.
Catalog No. 50-233-6788 Supplier Sepax Technologies Inc Supplier No. 272101P2105
Sepax offers an off-the-shelf product range of affinity columns conjugated with Human Serum Albumin (HSA) for our customers who are looking to asset mAb Immunogen Cross-Reactivity. The columns available can be easily attached to standard HPLC/UPLC instruments and provide an unique solution for process monitoring. For customers interested in custom immobilization contact us for more information.