Learn More
Sepax Technologies Inc Sepax HPLC Column BR-C18, 3 µm, 120 Å, 4.6 x 50 mm
SDP
Supplier:  Sepax Technologies Inc 1021834605
Encompass
Utilizing highest purity and enhanced mechanical stability silica and pure bonding reagents, BR-C18 bonded phases have been innovatively and specially designed to ensure maximum surface coverage and full end-capping, which leads to carbon content as high as 19.0%. The bonding chemistry is completely controlled that results in very reliable column-to-column reproducibility. The maximum surface coverage allows BR-C18 to have exceptional stability, resulting in high pH stability in the range of 1.5 to 10.5.
Catalog No. 50-233-6747