Learn More
Sepax Technologies Inc Sepax HPLC Column Carbomix K-NP10, 5% Crosslinkage, 10 x 300 mm
SDP
Catalog No. 502337595
Encompass
Carbomix phases are composed of spherical, low cross-linking polystyrene/divinylbenzene (PS/DVB) particles with their surfaces modified with sulfonic acid (-SO3H), followed by chelating of metal ions of calcium (Ca2+), lead (Pb2+), sodium (Na+) or potassium (K+) to form Carbomix Ca-NP, Pb-NP, Na-NP, K-NP phases, respectively. The PS/DVB support has cross-linking degree of 5%, 8% and 10% with the particle sizes of 5 and 10 µm. The uniform particle size adds the benefit of high separation efficiency to Carbomix. Carbomix resins have been specifically designed for high resolution separation of carbohydrates, organic acids, peptides, and nucleic acids.
Catalog No. 50-233-7595 Supplier Sepax Technologies Inc Supplier No. 23100510030
Carbomix phases are composed of spherical, low cross-linking polystyrene/divinylbenzene (PS/DVB) particles with their surfaces modified with sulfonic acid (-SO3H), followed by chelating of metal ions of calcium (Ca2+), lead (Pb2+), sodium (Na+) or potassium (K+) to form Carbomix Ca-NP, Pb-NP, Na-NP, K-NP phases, respectively. The PS/DVB support has cross-linking degree of 5%, 8% and 10% with the particle sizes of 5 and 10 µm. The uniform particle size adds the benefit of high separation efficiency to Carbomix. Carbomix resins have been specifically designed for high resolution separation of carbohydrates, organic acids, peptides, and nucleic acids.