Learn More
Sepax Technologies Inc Sepax HPLC Column GP-C18, 3 µm, 120 Å, 2.1 x 30 mm
SDP
Supplier:  Sepax Technologies Inc 1011832103
Encompass
GP-C18 uses full coverage bonded silica packing, which provides exceptionally high stability. The unique mono-functional bonding chemistry of GP-C18 avoids the formation of multiple C18 layers. Such uniform stationary phase allows the separation to achieve high selectivity and high efficiency.
Catalog No. 50-233-6593