Learn More
Sepax Technologies Inc Sepax HPLC Column GP-C18, 5 µm, 120 Å, 30 x 50 mm
SDP
Catalog No. 502336958
Encompass
GP-C18 uses full coverage bonded silica packing, which provides exceptionally high stability. The unique mono-functional bonding chemistry of GP-C18 avoids the formation of multiple C18 layers. Such uniform stationary phase allows the separation to achieve high selectivity and high efficiency.
Catalog No. 50-233-6958 Supplier Sepax Technologies Inc Supplier No. 10118530005
GP-C18 uses full coverage bonded silica packing, which provides exceptionally high stability. The unique mono-functional bonding chemistry of GP-C18 avoids the formation of multiple C18 layers. Such uniform stationary phase allows the separation to achieve high selectivity and high efficiency.