Learn More
Sepax Technologies Inc Sepax HPLC Column GP-C18, 5 µm, 120 Å, 4.6 x 50 mm
SDP
Catalog No. 502337515
Encompass
GP-C18 uses full coverage bonded silica packing, which provides exceptionally high stability. The unique mono-functional bonding chemistry of GP-C18 avoids the formation of multiple C18 layers. Such uniform stationary phase allows the separation to achieve high selectivity and high efficiency.
Catalog No. 50-233-7515 Supplier Sepax Technologies Inc Supplier No. 1011854605
Add to Cart
Add to Cart
GP-C18 uses full coverage bonded silica packing, which provides exceptionally high stability. The unique mono-functional bonding chemistry of GP-C18 avoids the formation of multiple C18 layers. Such uniform stationary phase allows the separation to achieve high selectivity and high efficiency.