Learn More
Sepax Technologies Inc Sepax HPLC Column GP-C4, 1.8 µm, 120 Å, 4.6 x 50 mm
SDP
Catalog No. 502337819
Encompass
Monomeric and fully endcapped GP-C4 packing is bonded with butyl groups that lead to moderate hydrophobicity. GP-C4 columns have unique selectivity and peak symmetry with moderate retention for separations of acidic, neutral and basic organic compounds, such as pharmaceuticals, peptides, and organic acids.
Catalog No. 50-233-7819 Supplier Sepax Technologies Inc Supplier No. 1090414605
Add to Cart
Add to Cart
Monomeric and fully endcapped GP-C4 packing is bonded with butyl groups that lead to moderate hydrophobicity. GP-C4 columns have unique selectivity and peak symmetry with moderate retention for separations of acidic, neutral and basic organic compounds, such as pharmaceuticals, peptides, and organic acids.