Learn More
Sepax Technologies Inc Sepax HPLC Column GP-C4, 5 µm, 120 Å, 30 x 150 mm
SDP
Catalog No. 502336664
Encompass
Monomeric and fully endcapped GP-C4 packing is bonded with butyl groups that lead to moderate hydrophobicity. GP-C4 columns have unique selectivity and peak symmetry with moderate retention for separations of acidic, neutral and basic organic compounds, such as pharmaceuticals, peptides, and organic acids.
Catalog No. 50-233-6664 Supplier Sepax Technologies Inc Supplier No. 10904530015
Add to Cart
Add to Cart
Request a Quote
Monomeric and fully endcapped GP-C4 packing is bonded with butyl groups that lead to moderate hydrophobicity. GP-C4 columns have unique selectivity and peak symmetry with moderate retention for separations of acidic, neutral and basic organic compounds, such as pharmaceuticals, peptides, and organic acids.