Learn More
Sepax Technologies Inc Sepax HPLC Column GP-C4, Guard cartridge with holder, 5 µm, 120 Å, 2 x 10 mm
SDP
Catalog No. 502337029
Encompass
Monomeric and fully endcapped GP-C4 packing is bonded with butyl groups that lead to moderate hydrophobicity. GP-C4 columns have unique selectivity and peak symmetry with moderate retention for separations of acidic, neutral and basic organic compounds, such as pharmaceuticals, peptides, and organic acids.
Catalog No. 50-233-7029 Supplier Sepax Technologies Inc Supplier No. 1090452001C
Monomeric and fully endcapped GP-C4 packing is bonded with butyl groups that lead to moderate hydrophobicity. GP-C4 columns have unique selectivity and peak symmetry with moderate retention for separations of acidic, neutral and basic organic compounds, such as pharmaceuticals, peptides, and organic acids.