Learn More
Sepax Technologies Inc Sepax HPLC Column GP-Phenyl, Guard cartridge with holder, 5 µm, 120 Å, 4 x 10 mm
SDP
Catalog No. 502338239
Encompass
GP-Phenyl packing materials are bonded with propyl phenyl groups that enable special interaction with ring structured compounds. The monomeric bonding chemistry gives high efficiency and high resolution separations. GP-Phenyl phase is suitable for separations of acidic, neutral and basic organic compounds, as well as pharmaceuticals.
Catalog No. 50-233-8239 Supplier Sepax Technologies Inc Supplier No. 1113654001C
GP-Phenyl packing materials are bonded with propyl phenyl groups that enable special interaction with ring structured compounds. The monomeric bonding chemistry gives high efficiency and high resolution separations. GP-Phenyl phase is suitable for separations of acidic, neutral and basic organic compounds, as well as pharmaceuticals.