Learn More
Sepax Technologies Inc Sepax HPLC Column HP-C18, 3 µm, 200 Å, 4.6 x 50 mm
SDP
Supplier:  Sepax Technologies Inc 1041834605
Encompass
HP-C18 uses full coverage bonded silica packing, which provides high stability. Compatible with 100% aqueous mobile phase and suitable for separations of acidic, neutral and basic organic compounds, as well as pharmaceuticals and peptides. ODS monolayer formed by special bonding chemistry does not collapse in pure aqueous solution.
Catalog No. 50-233-8499