Learn More
Sepax Technologies Inc Sepax HPLC Column HP-C18, 5 µm, 200 Å, 30 x 150 mm
SDP
Catalog No. 502337635
Encompass
HP-C18 uses full coverage bonded silica packing, which provides high stability. Compatible with 100% aqueous mobile phase and suitable for separations of acidic, neutral and basic organic compounds, as well as pharmaceuticals and peptides. ODS monolayer formed by special bonding chemistry does not collapse in pure aqueous solution.
Catalog No. 50-233-7635 Supplier Sepax Technologies Inc Supplier No. 10418530015
Add to Cart
Add to Cart
Request a Quote
HP-C18 uses full coverage bonded silica packing, which provides high stability. Compatible with 100% aqueous mobile phase and suitable for separations of acidic, neutral and basic organic compounds, as well as pharmaceuticals and peptides. ODS monolayer formed by special bonding chemistry does not collapse in pure aqueous solution.