Learn More
Sepax Technologies Inc Sepax HPLC Column HP-Cyano, 10 µm, 120 Å, 21.2 x 50 mm
SDP
Supplier:  Sepax Technologies Inc 11331921205
Encompass
Synthesized with monomeric and fully endcapped chemistry, HP-Cyano phase is bonded with propylcyano functional group that allows special interaction with polar compounds. The monomeric bonding chemistry enables high efficiency and high resolution separation of peptide, proteins, acidic, neutral and basic organic compound, and pharmaceuticals.
Catalog No. 50-233-7679