Learn More
Sepax Technologies Inc Sepax HPLC Column HP-Cyano, 3 µm, 120 Å, 2.1 x 250 mm
SDP
Catalog No. 502336267
Encompass
Synthesized with monomeric and fully endcapped chemistry, HP-Cyano phase is bonded with propylcyano functional group that allows special interaction with polar compounds. The monomeric bonding chemistry enables high efficiency and high resolution separation of peptide, proteins, acidic, neutral and basic organic compound, and pharmaceuticals.
Catalog No. 50-233-6267 Supplier Sepax Technologies Inc Supplier No. 1133132125
Add to Cart
Add to Cart
Synthesized with monomeric and fully endcapped chemistry, HP-Cyano phase is bonded with propylcyano functional group that allows special interaction with polar compounds. The monomeric bonding chemistry enables high efficiency and high resolution separation of peptide, proteins, acidic, neutral and basic organic compound, and pharmaceuticals.