Learn More
Sepax Technologies Inc Sepax HPLC Column HP-Cyano, 5 µm, 120 Å, 4.6 x 150 mm
SDP
Supplier:  Sepax Technologies Inc 1133154615
Encompass
Synthesized with monomeric and fully endcapped chemistry, HP-Cyano phase is bonded with propylcyano functional group that allows special interaction with polar compounds. The monomeric bonding chemistry enables high efficiency and high resolution separation of peptide, proteins, acidic, neutral and basic organic compound, and pharmaceuticals.
Catalog No. 50-233-8723