Learn More
Sepax Technologies Inc Sepax HPLC Column HP-Diol, 3 µm, 120 Å, 4.6 x 50 mm
SDP
Catalog No. 502338206
Encompass
Synthesized with polymeric chemistry, HP-Diol phase is bonded with 1,2-dihydroxypropyl propyl ether functional group that allows special interaction with polar compounds. The polymeric bonding chemistry enables high efficiency and high stability separation of polar pharmaceuticals, peptide, and proteins. HP-Diol phase can also be used as size exclusion separation of biological molecules.
Catalog No. 50-233-8206 Supplier Sepax Technologies Inc Supplier No. 1164234605
Synthesized with polymeric chemistry, HP-Diol phase is bonded with 1,2-dihydroxypropyl propyl ether functional group that allows special interaction with polar compounds. The polymeric bonding chemistry enables high efficiency and high stability separation of polar pharmaceuticals, peptide, and proteins. HP-Diol phase can also be used as size exclusion separation of biological molecules.