Learn More
Sepax Technologies Inc Sepax HPLC Column HPLC Guard, GP-Phenyl, 1.8 µm, 120 Å, 2.0 × 10 mm, Guard Cartridge with Holder
SDP
Catalog No. 502338285
Encompass
GP-Phenyl packing materials are bonded with propyl phenyl groups that enable special interaction with ring structured compounds. The monomeric bonding chemistry gives high efficiency and high resolution separations. GP-Phenyl phase is suitable for separations of acidic, neutral and basic organic compounds, as well as pharmaceuticals.
Catalog No. 50-233-8285 Supplier Sepax Technologies Inc Supplier No. 1113612001C
GP-Phenyl packing materials are bonded with propyl phenyl groups that enable special interaction with ring structured compounds. The monomeric bonding chemistry gives high efficiency and high resolution separations. GP-Phenyl phase is suitable for separations of acidic, neutral and basic organic compounds, as well as pharmaceuticals.