Learn More
Sepax Technologies Inc Sepax HPLC Column HPLC Guard Column, Zenix SEC-300, 3 µm, 7.8 x 50 mm, Light Scattering SEC
SDP
Catalog No. 502337177
Encompass
Made of 3 µm silica particles, Zenix SEC packing is a breakthrough technology for size exclusion chromatography. Combination of 3 µm silica particle size and proprietary surface technology enables highest separation efficiency and resolution for biological molecules and water soluble polymers. Available pore sizes of Zenix packings are 80, 100, 150 and 300 Å
Catalog No. 50-233-7177 Supplier Sepax Technologies Inc Supplier No. 213300LS7805
Made of 3 µm silica particles, Zenix SEC packing is a breakthrough technology for size exclusion chromatography. Combination of 3 µm silica particle size and proprietary surface technology enables highest separation efficiency and resolution for biological molecules and water soluble polymers. Available pore sizes of Zenix packings are 80, 100, 150 and 300 Å