Learn More
Sepax Technologies Inc Sepax HPLC Column HPLC GUARD Column, BioMix SEC-300, 3 µm, 4.6 x 50 mm
SDP
Catalog No. 502338673
Encompass
Utilizing proprietary surface technologies and 3 µm particle size, BioMix-300 phase is made of uniform, hydrophilic, and neutral nanometer thick films chemically bonded on the high purity and enhanced mechanical stability silica. The proprietary surface technologies allow the chemistry of thin film formation to be well controlled, which results in high column-to-column reproducibility. The nature of the chemical bonding and the uniformity of the thin film benefit BioMix-300 phase with high stability. BioMix-300 resin is produced from 3 µm spherical silica particles with a pore size of 300 Å. BioMix-300 columns are packed with a proprietary slurry technique to achieve uniform and stable packing bed density for maximum column efficiency. BioMix-300 phase is designed to ensure highest resolution and maximum recovery for a broad range of analytes such as bispecific mAb, hydrophobic samples etc.
Catalog No. 50-233-8673 Supplier Sepax Technologies Inc Supplier No. 2143004605
Utilizing proprietary surface technologies and 3 µm particle size, BioMix-300 phase is made of uniform, hydrophilic, and neutral nanometer thick films chemically bonded on the high purity and enhanced mechanical stability silica. The proprietary surface technologies allow the chemistry of thin film formation to be well controlled, which results in high column-to-column reproducibility. The nature of the chemical bonding and the uniformity of the thin film benefit BioMix-300 phase with high stability. BioMix-300 resin is produced from 3 µm spherical silica particles with a pore size of 300 Å. BioMix-300 columns are packed with a proprietary slurry technique to achieve uniform and stable packing bed density for maximum column efficiency. BioMix-300 phase is designed to ensure highest resolution and maximum recovery for a broad range of analytes such as bispecific mAb, hydrophobic samples etc.