Learn More
Sepax Technologies Inc Sepax HPLC Column HP-Silica, 10 µm, 120 Å, 21.2 x 100 mm
SDP
Catalog No. 502336282
Encompass
HP-Silica phase is made of activated hydroxyl (-OH) functional group with the pore size selection of 120 Å and particle size selection of 1.8, 3, 5, and 10 µm. Carbon loading is 0.0%. HP-Silica is used as the normal phase as well as HILIC phase. HP-Silica phases are suitable for separations of polar and basic organic compounds, such as vitamins, steroids, as well as pharmaceuticals.
Catalog No. 50-233-6282 Supplier Sepax Technologies Inc Supplier No. 11700921210
HP-Silica phase is made of activated hydroxyl (-OH) functional group with the pore size selection of 120 Å and particle size selection of 1.8, 3, 5, and 10 µm. Carbon loading is 0.0%. HP-Silica is used as the normal phase as well as HILIC phase. HP-Silica phases are suitable for separations of polar and basic organic compounds, such as vitamins, steroids, as well as pharmaceuticals.