Learn More
Sepax Technologies Inc Sepax HPLC Column Monomix MC10-SEC, 10 µm, 1000 Å, 7.8 x 300 mm
SDP
Supplier:  Sepax Technologies Inc 2801109507830
Encompass
Monomix MC SEC bulk media are highly crosslinked spherical resins made of polymethylacrylate. These rigid resins are narrowly dispersed particles with particle size selection of 10 μm and pore size selection of 500 and 1000 Å. These resins have proprietary hydrophilic surface functional groups that minimize non-specific binding. Monomix MC SEC resins possess several key benefits: a broad pH tolerant range (1-14), elevated operating temperature (up to 80°C), high operating pressure (up to 20 bar), monosized particle (D90/D10 < 1.3, low column back pressure, high theoretical plate number), smooth surface and highly spherical (for easy column packing).
Catalog No. 50-233-7034