Learn More
Sepax Technologies Inc Sepax HPLC Column Polar-Imidazole, 3 µm, 120 Å, 2 x 10 mm
SDP
Catalog No. 502336932
Encompass
Utilizing highest purity and enhanced mechanical stability silica and pure bonding reagents, Polar-Imidazole phase is bonded with imidazole functional groups. With a polar and N-containing ring structure, Polar-Pyridine phase is a weak base and adequate for many applications, such as pharmaceuticals, peptides, sugars, and nucleotides.
Catalog No. 50-233-6932 Supplier Sepax Technologies Inc Supplier No. 1353332001
Add to Cart
Add to Cart
Utilizing highest purity and enhanced mechanical stability silica and pure bonding reagents, Polar-Imidazole phase is bonded with imidazole functional groups. With a polar and N-containing ring structure, Polar-Pyridine phase is a weak base and adequate for many applications, such as pharmaceuticals, peptides, sugars, and nucleotides.