Learn More
Sepax Technologies Inc Sepax HPLC Column PolyRP-NP5, Guard cartridge with holder, 4 x 10 mm
SDP
Supplier:  Sepax Technologies Inc 2620054001C
Encompass
GP-C8 phase is synthesized with monomeric and fully endcapped chemistry. The uniform octyl stationary phase allows high efficiency with lower hydrophobicity compared to ODS phase. GP-C8 phase is suitable for separating compounds with a wide range of hydrophobicity. It is highly recommended for separating the compounds which are too strongly retained on C18 phases.
Catalog No. 50-233-6529