Learn More
Sepax Technologies Inc Sepax HPLC Column Proteomix HIC Butyl-NP2, 4 x 10 mm
SDP
Catalog No. 502337562
Encompass
Proteomix HIC columns are specially designed for high resolution and high efficiency separations of proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs), oligonucleotides and peptides via a hydrophobic interaction chromatography (HIC) mechanism. Utilizing proprietary surface technologies, Proteomix HIC-NP resin is made of non-porous polystyrenedivinylbenzene (PS/DVB) beads with narrow-dispersed particle size distribution. As shown in figure below, the PS/DVB bead is modified with alkyl groups or aryl groups that provide hydrophobic interaction with analytes. Proteomix HIC-NP resin is highly rigid and mechanically stable. In comparison to silica-based HIC phase media, Proteomix HIC-NP phases have advantages for biomolecule separations with a wide pH range (2-12) and high chemical stability.
Catalog No. 50-233-7562 Supplier Sepax Technologies Inc Supplier No. 431NP24001
Proteomix HIC columns are specially designed for high resolution and high efficiency separations of proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs), oligonucleotides and peptides via a hydrophobic interaction chromatography (HIC) mechanism. Utilizing proprietary surface technologies, Proteomix HIC-NP resin is made of non-porous polystyrenedivinylbenzene (PS/DVB) beads with narrow-dispersed particle size distribution. As shown in figure below, the PS/DVB bead is modified with alkyl groups or aryl groups that provide hydrophobic interaction with analytes. Proteomix HIC-NP resin is highly rigid and mechanically stable. In comparison to silica-based HIC phase media, Proteomix HIC-NP phases have advantages for biomolecule separations with a wide pH range (2-12) and high chemical stability.