Learn More
Sepax Technologies Inc Sepax HPLC Column Proteomix WAX, NP5 PEEK, 2.1 x 250 mm
SDP
Supplier:  Sepax Technologies Inc 404NP5P2125
Encompass
Sepax HPLC Column Proteomix WAX, NP5 PEEK, 2.1 x 250 mm
Catalog No. 50-233-8158