Learn More
Sepax Technologies Inc Sepax HPLC Column TweenTrap, 5 µm, NP, 4.6 x 35 mm
SDP
Catalog No. 502337544
Encompass
TweenTrap columns are specially designed for trapping surfactants, such as Tweens, in aqueous samples. Their packing support is composed of rigid, spherical, highly cross-linked poly(styrene divinylbenzene) (PS/DVB) beads with a particle size of 5 µm. A proprietary surface coating is covalently bonded onto this PS/DVB support, which allows for exceptionally high stability. These columns are compatible with most aqueous buffers such as acetate, phosphate, and tris.
Catalog No. 50-233-7544 Supplier Sepax Technologies Inc Supplier No. 0100544603
TweenTrap columns are specially designed for trapping surfactants, such as Tweens, in aqueous samples. Their packing support is composed of rigid, spherical, highly cross-linked poly(styrene divinylbenzene) (PS/DVB) beads with a particle size of 5 µm. A proprietary surface coating is covalently bonded onto this PS/DVB support, which allows for exceptionally high stability. These columns are compatible with most aqueous buffers such as acetate, phosphate, and tris.