Learn More
Sepax Technologies Inc Sepax HPLC Column Unix™ SEC-300, 1.8 µm, 4.6 x 300 mm
SDP
Supplier:  Sepax Technologies Inc 2113004630
Encompass
Utilizing proprietary surface technologies, Unix SEC phase is made of uniform, hydrophilic, and neutral nanometer thick films chemically bonded to high purity and mechanically stabilized silica with the particle size of 1.8 µm. The combination of small particle size and large pore volume of Unix SEC renders the highest separation efficiency and resolution of analytes. The well-controlled surface chemistry results in excellent lot-to-lot reproducibility. Our unique bonding chemistry, coupled with the maximized bonding density, allows Unix SEC to provide high stability and negligible non-specific interactions. Typical applications for Unix SEC columns include separation and analysis of biological molecules and water-soluble polymers.
Catalog No. 50-233-7188