Learn More
Sepax Technologies Inc Sepax HPLC Column Unix™-C SEC-300, 1.8 µm, 4.6 x 300 mm
SDP
Catalog No. 502336358
Encompass
Utilizing proprietary innovative surface technologies, Unix™-C SEC-300 phase is made of uniform, hydrophilic, and neutral nanometer thick films chemically bonded to high purity and mechanically stabilized silica with the particle size of 1.8 µm. The combination of small particle size and large pore volume of Unix™-C SEC-300 renders the highest separation efficiency and resolution of analytes. The well-controlled surface chemistry results in excellent lot-to-lot reproducibility. The unique bonding chemistry, coupled with the maximized bonding density, allows Unix™-C SEC-300 to provide high stability and negligible non-specific interactions. Unix™-C, uses lay-down monolayer on porous silica, offers ideal phase chemistry for the separation of all biological sample types including challenging hydrophobic samples
Catalog No. 50-233-6358 Supplier Sepax Technologies Inc Supplier No. 2313004630
Utilizing proprietary innovative surface technologies, Unix™-C SEC-300 phase is made of uniform, hydrophilic, and neutral nanometer thick films chemically bonded to high purity and mechanically stabilized silica with the particle size of 1.8 µm. The combination of small particle size and large pore volume of Unix™-C SEC-300 renders the highest separation efficiency and resolution of analytes. The well-controlled surface chemistry results in excellent lot-to-lot reproducibility. The unique bonding chemistry, coupled with the maximized bonding density, allows Unix™-C SEC-300 to provide high stability and negligible non-specific interactions. Unix™-C, uses lay-down monolayer on porous silica, offers ideal phase chemistry for the separation of all biological sample types including challenging hydrophobic samples