Learn More
Sepax Technologies Inc Sepax Protein standard for Proteomix SCX and WCX (5 vials, 50ul/vial) -Ribonuclease A (1 mg/mL) -Cytochrome c (1 mg/mL) -Lysozyme (1 mg/mL)
SDP
Catalog No. 502336943
Encompass
Protein standards are a group of proteins that are used as the standards to calibrate various modes of separation phases listed below. Those standards are supplied as a set of 5 vials of protein mixture.
Catalog No. 50-233-6943 Supplier Sepax Technologies Inc Supplier No. 2010040000
Protein standards are a group of proteins that are used as the standards to calibrate various modes of separation phases listed below. Those standards are supplied as a set of 5 vials of protein mixture.