Learn More
Sepax Technologies Inc Sepax Protein standard for Proteomix HIC (5 vials, 50ul/vial) -Ovalbumin (1 mg/mL) -Chymotrypsinogen (0.5 mg/mL)
SDP
Catalog No. 502336624
Encompass
Protein standards are a group of proteins that are used as the standards to calibrate various modes of separation phases listed below. Those standards are supplied as a set of 5 vials of protein mixture.
Catalog No. 50-233-6624 Supplier Sepax Technologies Inc Supplier No. 2010070000
Protein standards are a group of proteins that are used as the standards to calibrate various modes of separation phases listed below. Those standards are supplied as a set of 5 vials of protein mixture.